missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Osteoprotegerin/TNFRSF11B Protein

Product Code. 18178450
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18178450

Brand: R&D Systems™ 185OS025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A New and Improved rh OPG is Now Available! The new protein has ∼2 fold better activity!

Measured by its ability to inhibit TRAIL-mediated cytotoxicity using L-929 mouse fibroblast cells treated with TRAIL. The ED50 for this effect is 8-24 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAB53709.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 4982
Molecular Weight (g/mol) M.W. (Observed): 55 kDa, reducing conditions; M.W. (theoretical): 43.5 kDa (monomer)
Name Osteoprotegerin/TNFRSF11B
Quantity 25 μg
Source Mouse myeloma cell line, NS0-derived human Osteoprotegerin/TNFRSF11B protein Glu22-Leu401
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol TNFRSF11B
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Osteoprotegerin/TNFRSF11B
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.