missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Netrin-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£399.56
Specifications
Gene ID (Entrez) | 9423 |
---|---|
Molecular Weight (g/mol) | 67.5 kDa |
Name | Netrin-1 |
Quantity | 25μg |
Source | Mouse myeloma cell line,NS0-derived human Netrin-1 protein, (N-terminus) Human Netrin-1 (Val22-Ala604) Accession &Num; O95631 HPGGGSGGGSGGGS 6-His tag (C-terminus) |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18368977
|
R&D Systems™
6419-N1-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS and EDTA. |
£399.56
25µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18187240
|
R&D Systems™
6419-N1-025 |
Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein. |
£399.56
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human Netrin-1 Protein is derived from NS0. The Recombinant Human Netrin-1 Protein has been validated for the following applications: Bioactivity.Specifications
9423 | |
Netrin-1 | |
Mouse myeloma cell line,NS0-derived human Netrin-1 protein, (N-terminus) Human Netrin-1 (Val22-Ala604) Accession &Num; O95631 HPGGGSGGGSGGGS 6-His tag (C-terminus) | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
67.5 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |