missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human MSP/MST1 β Chain Recombinant Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 4485 |
Molecular Weight (g/mol) | 25 kDa |
Quantity | 25μg |
Recombinant | Yes |
Description
The Recombinant Human MSP/MST1 beta Chain Protein is derived from NS0. The Recombinant Human MSP/MST1 beta Chain Protein has been validated for the following applications: Binding Activity.Specifications
Unconjugated | |
25 kDa | |
Yes |
4485 | |
25μg | |
Mouse myeloma cell line,NS0-derived human MSP/MST1 protein Val484-Gly711 |