missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human MS4A6A (NP_690591) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Beschreibung
The Human MS4A6A (NP_690591) VersaClone cDNA clone contains the complete ORF for MS4A6A, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The MS4A6A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Bestellinformation
Ships at ambient temperature.
Spezifikation
Spezifikation
| Gene | hMS4A6A |
| Accession Number | NP_690591 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Insert Type | 759bp |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur