missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Meprin alpha Subunit/MEP1A Protein, CF

Product Code. 18359207
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359207 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359207 Supplier R&D Systems™ Supplier No. 3220ZN010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave a fluorogenic peptide substrate, Mca-YVADAPK(Dnp)-OH (Catalog # ES007 ). The specific activity is >500 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAA21338
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 4224
Molecular Weight (g/mol) M.W. (Observed): 86 kDa, reducing conditions; M.W. (theoretical): 67 kDa
Name Meprin alpha Subunit/MEP1A
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived human Meprin alpha Subunit/MEP1A protein Val22-Gln601 & Leu27-Gln601, both with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol MEP1A
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Meprin alpha Subunit/MEP1A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.