missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human LIGHT/TNFSF14 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Unit Size:
25µg
Description
Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 for this effect is 0.3-3 ng/mL.
Specifications
Specifications
| Accession Number | O43557 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 8740 |
| Molecular Weight (g/mol) | M.W. (Observed): 25 kDa, reducing conditions; M.W. (theoretical): 20.9 kDa |
| Name | LIGHT/TNFSF14 |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line, NS0-derived human LIGHT/TNFSF14 protein Met 10-His tag GGGSGGGSGGGSIEGR Human Light (Asp74-Val240) (N-terminus) Accession # O43557 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction