missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-7R alpha/CD127 Fc Chimera Protein, CF
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human IL-7 R alpha/CD127 Fc Chimera (0.25 μg/mL, 100 μL/well) is immobilized on a goat anti-human IgG Fc antibody (Catalog # G-102-C ) coated plate, the concentration of recombinant human IL-7 that produces 50% of the optimal binding response is found to be 7.5-60 ng/mL.
Specifications
Specifications
| Accession Number | BAG62793.1 |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 3575 |
| Molecular Weight (g/mol) | 54.5 kDa (monomer) |
| Name | IL-7R alpha/CD127 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human IL-7 R alpha/CD127 protein, (N-terminus) Human IL-7 R alpha (Glu21-Lys261) Accession &Num; BAG62793.1 IIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction