missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-36 gamma/IL-1F9 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
£278.61
Specifications
Gene ID (Entrez) | 56300 |
---|---|
Molecular Weight (g/mol) | 18.7 kDa |
Name | IL-36 gamma/IL-1F9 |
Quantity | 25μg |
Source | E. coli-derived human IL-36 gamma/IL-1F9 protein Met1-Asp169 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18147010
|
R&D Systems™
2320-IL-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£278.61
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18197060
|
R&D Systems™
2320-IL-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£278.61
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human IL-36 gamma/IL-1F9 Protein is derived from E. coli. The Recombinant Human IL-36 gamma/IL-1F9 Protein has been validated for the following applications: Binding Activity.Specifications
56300 | |
IL-36 gamma/IL-1F9 | |
E. coli-derived human IL-36 gamma/IL-1F9 protein Met1-Asp169 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
18.7 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |