missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-2 Protein

Product Code. 18370328
Change view
Click to view available options
Quantity:
10 μg
50 μg
500 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
Unit Size:
10µg
500µg
50µg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Formulation unitSize
18370328 50 μg Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA. 50µg
18116540 10 μg Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA. 10µg
18374018 10 μg Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein. 10µg
18157790 50 μg Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein. 50µg
18624749 500 μg Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein. 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 18370328 Supplier R&D Systems™ Supplier No. 202IL050/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Version with mutation found in aldesleukin Available!

Measured in a cell proliferation assay using CTLL-2 mouse cytotoxic T cells. Gearing, A.J.H. and C.B. Bird (1987) in Lymphokines and Interferons, A Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 295. The ED50 for this effect is 0.05-0.25 ng/mL. The specific activity of Recombinant Human IL-2 is approximately 2.1 x 104 IU/μg, which is calibrated against recombinant human IL-2 WHO International Standard (NIBSC code: 86/500).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P60568
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 3558
Molecular Weight (g/mol) 13 kDa, under reducing conditions.
Name IL-2
Quantity 50 μg
Source E. coli-derived human IL-2 protein Ala21-Thr153, with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol IL2
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile 100 mM Acetic Acid. Alternatively, reconstitute at 100 μg/mL in sterile deionized water and use within 24 hours, store (at 2°C to 8°C.)
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein IL-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.