missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-17E Protein

Product Code. 18311678
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Unit Size:
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18311678 Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein. 25µg
18157770 Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18311678 Supplier R&D Systems™ Supplier No. 1258IL025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A New and Improved rh IL-17E is Now Available! The new protein is mammalian cell expressed!

Measured by its ability to induce CXCL1/GRO alpha secretion in HT-29 human colon adenocarcinoma cells. The ED50 for this effect is 0.25-1.5 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H293
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 64806
Molecular Weight (g/mol) M.W. (theoretical): 17 kDa (monomer)
Name IL-17E/IL-25
Quantity 25 μg
Source E. coli-derived human IL-17 E/IL-25 protein Tyr33-Gly177, with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol IL25
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile 4 mM HCl containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein IL-17 E/IL-25
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.