missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-12 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£339.08 - £896.30
Specifications
Gene ID (Entrez) | 3592 |
---|---|
Molecular Weight (g/mol) | 34.7 kDa (p40) & 22.5 kDa (p35) |
Name | IL-12 |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-12 protein, (N-terminus) Human IL-12 p40 (Ile23-Ser328) Accession &Num; P29460 Human IL-12 p35 (Arg23-Ser219) Accession &Num; P29459 (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
18325007
|
R&D Systems™
219-IL-005/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 5μg |
£339.08
5µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18358947
|
R&D Systems™
219-IL-005 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 5μg |
£339.08
5µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18166780
|
R&D Systems™
219-IL-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 25μg |
£896.30
25µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18106650
|
R&D Systems™
219-IL-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 25μg |
£896.30
25µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Measured in a cell proliferation assay using PHA-stimulated human T lymphoblasts. Symons, J.A. et al. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 272. The ED50 for this effect is 0.01-0.05 ng/mL.Specifications
3592 | |
IL-12 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE with silver staining. |
34.7 kDa (p40) & 22.5 kDa (p35) | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-12 protein, (N-terminus) Human IL-12 p40 (Ile23-Ser328) Accession &Num; P29460 Human IL-12 p35 (Arg23-Ser219) Accession &Num; P29459 (C-terminus) | |
Disulfide-linked heterodimer | |
Recombinant |