missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IFN-alpha A/D [BglII] (Universal Type I Interferon) Recombinant Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Specifications
| Formulation | Phosphate buffered saline (PBS) containing 0.1% bovine serum albumin (BSA) |
|---|---|
| Purification Method | Purified from E. coli by a combination of hydrophobic interaction, ion exchange and gel permeation chromatography |
| Recombinant | Yes |
Description
IgG Polyclonal antibody specifically detects IgG in Human samples. It is validated for Control.Specifications
| Phosphate buffered saline (PBS) containing 0.1% bovine serum albumin (BSA) | |
| Purified from E. coli by a combination of hydrophobic interaction, ion exchange and gel permeation chromatography | |
| Yes |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.