missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Galectin-3 Protein

Product Code. 18117060
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS and Betamercaptoethanol with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS and Betamercaptoethanol.
Unit Size:
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18117060 Lyophilized from a 0.2 μm filtered solution in PBS and Betamercaptoethanol with BSA as a carrier protein. 50µg
18118400 Lyophilized from a 0.2 μm filtered solution in PBS and Betamercaptoethanol. 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18117060 Supplier R&D Systems™ Supplier No. 1154GA050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A New rh Galectin-3 is Now Available! Improved activity and purity; Mammalian cell expressed!

Measured by its ability to agglutinate human red blood cells. Hadari, Y.R. et al. (2000) J. Cell Sci. 113:2385. The ED50 for this effect is 3-10 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P17931
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and Betamercaptoethanol with BSA as a carrier protein.
Gene ID (Entrez) 3958
Molecular Weight (g/mol) M.W. (theoretical): 26 kDa
Name Galectin-3
Quantity 50 μg
Source E. coli-derived human Galectin-3 protein Ala2-Ile250
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 3 months, 2°C to 8°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol LGALS3
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 50 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Galectin-3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.