missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human EGFR Fc Chimera Protein, CF

Product Code. 18117230
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18117230 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18117230 Supplier R&D Systems™ Supplier No. 344ER050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its ability to bind recombinant human EGF in a functional ELISA with an estimated Kd <8 nM.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number CAA25240
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 1956
Molecular Weight (g/mol) M.W. (Observed): 125-145 kDa, reducing conditions; M.W. (theoretical): 95.1 kDa (monomer)
Name EGF R/ErbB1 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human EGFR protein Human EGFR (Met1-Ser645) (N-terminus) Accession # CAA25240 IEGRMD Human IgG1-Fc (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol EGFR
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein EGFR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.