missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CXCL5/ENA-70 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£228.94
Specifications
Gene ID (Entrez) | 6374 |
---|---|
Molecular Weight (g/mol) | 7.7 kDa |
Name | CXCL5/ENA-70 |
Quantity | 25μg |
Source | E. coli-derived human CXCL5/ENA-70 protein Arg45-Asn114 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18156370
|
R&D Systems™
649-EN-025/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
£228.94
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18137480
|
R&D Systems™
649-EN-025 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
£228.94
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human CXCL5/ENA-70 Protein is derived from E. coli. The Recombinant Human CXCL5/ENA-70 Protein has been validated for the following applications: Bioactivity.Specifications
6374 | |
CXCL5/ENA-70 | |
E. coli-derived human CXCL5/ENA-70 protein Arg45-Asn114 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
7.7 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |