missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CD40 Ligand/TNFSF5 (aa 108-261) Protein

Product Code. p-4719305
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and EDTA.
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18167060

Brand: R&D Systems™ 6245CL050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A New rh CD40 Ligand is Available! ∼1000 fold better activity; HEK293 expressed; HA tag!

Measured in a cell proliferation assay using B cell-enriched peripheral blood mononuclear cells (PBMC) in the presence of IL-4. Spriggs, M.K. et al. (1992) J. Exp. Med. 176:1543. The ED50 for this effect is 1-3.00 μg/mL in the presence of 20 ng/mL of recombinant human IL-4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29965
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and EDTA with BSA as a carrier protein.
Gene ID (Entrez) 959
Molecular Weight (g/mol) M.W. (Observed): 18 kDa, reducing conditions; M.W. (theoretical): 16.9 kDa (monomer)
Name CD40 Ligand/TNFSF5 aa 108-261
Quantity 50 μg
Source E. coli-derived human CD40 Ligand/TNFSF5 protein Glu108-Leu261, with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 3 months, 2°C to 8°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol CD40LG
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein CD40 Ligand/TNFSF5
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.