missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human/Cynomolgus/Rhesus Macaque CD28 Fc, CF

Product Code. 18355597
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355597 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355597 Supplier R&D Systems™ Supplier No. 342CD200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Human/Cynomolgus Monkey/Rhesus Macaque CD28 Fc Chimera is coated at 1 μg/mL (100 μL/well), the concentration of Recombinant Human B7-1/CD80 Fc Chimera (Catalog # 140-B1 ) that produces 50% of the optimal binding response is approximately 0.05-0.3 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P10747
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 940
Molecular Weight (g/mol) M.W. (Observed): 60-70 kDa, reducing conditions; M.W. (theoretical): 42.2 kDa (monomer)
Name CD28 Fc Chimera
Quantity 200 μg
Source Mouse myeloma cell line, NS0-derived CD28 protein Human/Cynomolgus Monkey/Rhesus Macaque CD28 (Asn19-Pro152) (N-terminus) Accession # P10747 DMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol CD28
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CD28
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.