missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CCL2/MCP-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£260.25 - £516.19
Specifications
Gene ID (Entrez) | 6347 |
---|---|
Molecular Weight (g/mol) | 8.7 kDa |
Name | CCL2/JE/MCP-1 |
Source | E. coli-derived human CCL2/JE/MCP-1 protein Gln24-Thr99 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
18319237
|
R&D Systems™
279-MC-010 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 10μg |
£260.25
10µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18369257
|
R&D Systems™
279-MC-010/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 10μg |
£260.25
10µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18186910
|
R&D Systems™
279-MC-050 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 50μg |
£516.19
50µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18136640
|
R&D Systems™
279-MC-050/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 50μg |
£516.19
50µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The Recombinant Human CCL2/MCP-1 Protein is derived from E. coli. The Recombinant Human CCL2/MCP-1 Protein has been validated for the following applications: Bioactivity.Specifications
6347 | |
CCL2/JE/MCP-1 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
8.7 kDa | |
E. coli-derived human CCL2/JE/MCP-1 protein Gln24-Thr99 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |