missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human BTN2A2/Butyrophilin 2A2 (NP_008926) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Human BTN2A2/Butyrophilin 2A2 (NP_008926) VersaClone cDNA clone contains the complete ORF for BTN2A2/Butyrophilin 2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The BTN2A2/Butyrophilin 2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
| Gene | BTN2A2/Butyrophilin 2 |
| Accession Number | NP_008926 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Insert Type | 1585bp |
| Gene ID (Entrez) | 10385 |
| Gene Alias | BTF2, BTF2BT2.2 butyrophilin 2, BTN2, Butyrophilin 2, butyrophilin subfamily 2 member A2, butyrophilin, subfamily 2, member A2, FLJ41908 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction