missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human APRIL/TNFSF13 (HEK293-expressed) Protein

Product Code. 18369617
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl.
Unit Size:
10µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18369617 Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein. 10µg
18316387 Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl. 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18369617 Supplier R&D Systems™ Supplier No. 5860AP010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using anti-IgM stimulated mouse B cells. The ED50 for this effect is 5-25 ng/mL in the presence of goat anti-mouse IgM.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75888
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein.
Gene ID (Entrez) 8741
Molecular Weight (g/mol) M.W. (Observed): 26 kDa, reducing conditions; M.W. (theoretical): 21.7 kDa
Name APRIL/TNFSF13
Quantity 10 μg
Source Human embryonic kidney cell, HEK293-derived human APRIL/TNFSF13 protein HA (YPYDVPDYA) GCN4-IZ GGGSGGGSGGGS Human April (Ala105-Leu250) (N-terminus) Accession # O75888 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Noncovalently-linked homotrimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol TNFSF13
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein APRIL/TNFSF13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.