missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human APRIL/TNFSF13 (HEK293-expressed) Protein

Product Code. 18316387
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl.
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18316387

Brand: R&D Systems™ 5860AP010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using anti-IgM stimulated mouse B cells. The ED50 for this effect is 5-25 ng/mL in the presence of goat anti-mouse IgM.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75888
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris-HCl and NaCl.
Gene ID (Entrez) 8741
Molecular Weight (g/mol) M.W. (Observed): 26 kDa, reducing conditions; M.W. (theoretical): 21.7 kDa
Name APRIL/TNFSF13
Quantity 10 μg
Source Human embryonic kidney cell, HEK293-derived human APRIL/TNFSF13 protein HA (YPYDVPDYA) GCN4-IZ GGGSGGGSGGGS Human April (Ala105-Leu250) (N-terminus) Accession # O75888 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Noncovalently-linked homotrimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol TNFSF13
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein APRIL/TNFSF13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.