missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Apo-Transferrin Protein

Product Code. p-4719084
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 18349477

Brand: R&D Systems™ 3188AT100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a serum-free cell proliferation assay using CHO-K1 cells. The ED50 for this effect is <6.00 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in deionized water.
Name Apo-Transferrin
Quantity 100 mg
Source Human plasma-derived Apo-Transferrin protein The human plasma used for the isolation of this product were certified by the supplier to be negative for HIV I and II antibodies, Hepatitis C antibody and Hepatitis B antigen at the time of shipment. Human blood products should always be treated in accordance with universal handling precautions.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.