missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Airway Trypsin-like Protease/HAT, CF

Product Code. 18196330
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18196330

Brand: R&D Systems™ 2695SE010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave the fluorogenic peptide substrate, t-butoxycarbonyl-Phe-Ser-Arg-7-amino-4-methyl coumarin (Boc-FSR-AMC). The specific activity is >50,000 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60235
For Use With (Application) Enzyme Activity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 9407
Molecular Weight (g/mol) M.W. (Observed): 27 kDa and 18 kDa, reducing conditions; M.W. (theoretical): 25 kDa & 17 kDa
Name Airway Trypsin-like Protease/HAT
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived human Airway Trypsin-like Protease/HAT protein Ala42-Ile418 with an N-terminal 6-His tag The protein was purified, activated and further purified.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Active
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol TMPRSS11D
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile 25 mM Tris, 150 mM NaCl and 0.05% Brij-35, pH 7.5.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Airway Trypsin-like Protease/HAT/TMPRSS11D
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.