missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Activin B Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£218.14 - £566.94
Specifications
Gene ID (Entrez) | 3625 |
---|---|
Molecular Weight (g/mol) | 12.8 kDa (monomer) |
Name | Activin B |
Source | Chinese Hamster Ovary cell line,CHO-derived human Activin B protein Gly293-Ala407 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
18178050
|
R&D Systems™
659-AB-005 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | 5μg |
£218.14
5µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18166420
|
R&D Systems™
659-AB-005/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with Trehalose. | 5μg |
£218.14
5µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18368387
|
R&D Systems™
659-AB-025 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | 25μg |
£566.94
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18117880
|
R&D Systems™
659-AB-025/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with Trehalose. | 25μg |
£566.94
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The Recombinant Human Activin B Protein is derived from CHO. The Recombinant Human Activin B Protein has been validated for the following applications: Bioactivity.Specifications
3625 | |
Activin B | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
activin AB beta polypeptide, Activin beta-B chain, INHBB, inhibin beta B chain, inhibin, beta B, inhibin, beta B (activin AB beta polypeptide), Inhibin, beta-2 | |
Recombinant |
12.8 kDa (monomer) | |
Chinese Hamster Ovary cell line,CHO-derived human Activin B protein Gly293-Ala407 | |
Disulfide-linked homodimer | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |