missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human 20S Proteasome Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Human 20S Proteasome is able to degrade substrates in an ATP-independent manner. Reaction conditions will need to be optimized for each specific application. We recommend an initial Human 20S Proteasome concentration of 0.5-5 nM.
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES, NaCl and DTT. |
| Molecular Weight (g/mol) | 700 kDa |
| Quantity | 50 μg |
| Source | Human erythrocyte-derived 20 S Proteasome protein The Human 20 S Proteasome protein has been purified from human erythrocytes, which have been screened and are negative for hepatitis B surface antigen, antibodies to hepatitis C virus, HIV type 1 antigens, and antibodies to HIV type 1 and 2. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Product Type | 20S Proteasome |
| Conjugate | Unconjugated |
| Species | Human |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction