missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flow Cytometry Permeabilization/Wash Buffer I (1X), R&D Systems™
Click to view available options
Quantity:
250mL
Unit Size:
250mL
Description
- Designed for intracellular flow cytometry applications with nonconjugated or fluorochrome-conjugated antibodies
Applications:
Flow Cytometry
Specifications
Specifications
| Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
| Formulation | Sodium azide 0.05% |
| Quantity | 250mL |
| Stability | Upto 6 Months |
| Product Type | Permeabilization/Wash Buffer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction