missing translation for 'onlineSavingsMsg'
Learn More

Flow Cytometry Fixation Buffer (1X), R&D Systems™

Product Code. 18762464
Change view
Click to view available options
Quantity:
100mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18762464 100mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18762464 Supplier R&D Systems Supplier No. FC004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: Flow Cytometry

  • Designed for intracellular flow cytometry applications with nonconjugated or fluorochrome-conjugated antibodies

Applications:

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Formulation Formaldehyde 4%
Quantity 100mL
Stability Upto 6 Months
Product Type Fixation Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.