missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Feline Flt-3 Ligand/FLT3L Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£344.48
Specifications
Gene ID (Entrez) | 493796 |
---|---|
Molecular Weight (g/mol) | Observed M.W.: 25-33 kDa, under reducing conditions. |
Name | Flt-3 Ligand |
Quantity | 25μg |
Source | Mouse myeloma cell line,NS0-derived feline Flt-3 Ligand/FLT3L protein Ser27-Pro185,with a C-terminal 6-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18197600
|
R&D Systems™
2467-FL-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£344.48
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18198640
|
R&D Systems™
2467-FL-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£344.48
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Feline Flt-3 Ligand Protein is derived from NS0. The Recombinant Feline Flt-3 Ligand Protein has been validated for the following applications: Bioactivity.Specifications
493796 | |
Flt-3 Ligand | |
Mouse myeloma cell line,NS0-derived feline Flt-3 Ligand/FLT3L protein Ser27-Pro185,with a C-terminal 6-His tag | |
FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine | |
Recombinant |
Observed M.W.: 25-33 kDa, under reducing conditions. | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |