missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ESI 09

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ESI 09 is an EPAC inhibitor. Inhibits EPAC-mediated insulin release from pancreatic β cells. Selective for EPAC over PKA. ESI 09 decreases migration and invasion of AsPc-1 and PANC-1 pancreatic cancer cells in vitro.
Specifications
Specifications
| Chemical Name or Material | EPAC Inhibitors |
| CAS | 263707-16-0 |
| Synonym | α-[(2-(3-Chlorophenyl)hydrazinylidene]-5-(1,1-dimethylethyl)-β-oxo-3-isoxazolepropanenitrile |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in DMSO and to 20mM in ethanol |
| Molecular Formula | C16H15ClN4O2 |
| Formula Weight | 330.77 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Partager une correction de contenu