missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DSR 6434
GREENER_CHOICE

Product Code. 18790939
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18790939 50 mg 50mg
18731548 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18790939 Supplier Tocris Bioscience™ Supplier No. 4809/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent TLR7 agonist

DSR 6434 is a potent TLR7 agonist (EC50 = 7.2 nM). Activates several immune effector cells in vitro. Suppresses metastasis in vivo and enhances efficacy of radiation therapy in a model of colorectal carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DSR 6434
CAS 1059070-10-8
Synonym 6-Amino-2-(butylamino)-9-[[6-[2-(dimethylamino)ethoxy]-3-pyridinyl]methyl]-7,9-dihydro-8H-purin-8-one
Quantity 50 mg
Solubility Soluble to 50mM in 1eq. HCl and to 100mM in DMSO
Molecular Formula C19H28N8O2
Formula Weight 400.48
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.