missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cultrex RGF Basement Membrane Extract, Type 2

Product Code. 18746007
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 x 5 mL
Unit Size:
10mL
1mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18746007 1 mL 1mL
18776937 5 mL 5mL
18748977 2 x 5 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18746007 Supplier R&D Systems™ Supplier No. 353300102

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Robust Organoid Cultures - Reduced Growth Factor Basement Membrane Extract

Tube Formation Assay - BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. Gelling Assay - BME gels in less than 30 minutes at 37degreesC, and maintains the gelled form in culture medium for a minimum of 14 days at 37degreesC.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 8-12 mg/mL (by Lowry assay)
For Use With (Application) Bioactivity
Quantity 1 mL
Source Murine Engelbreth-Holm-Swarm (EHS) tumor.
Content And Storage Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.