missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cultrex Basement Membrane Extract, PathClear

Product Code. p-4788192
Click to view available options
Quantity:
1 mL
5 mL
2 x 5 mL
Unit Size:
10mL
1mL
5mL
This item is not returnable. View return policy

Product Code. 18402240

Brand: R&D Systems™ 343201001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: Bioactivity

Functional Assay: Tube formation - BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. Gelling Assay: BME gels in less than 30 minutes at 37degreesC, and maintains the gelled form in culture medium for a minimum of 14 days at 37degreesC.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 12 to 18 mg/mL
For Use With (Application) Bioactivity
Quantity 2 x 5 mL
Endotoxin Level ≤8EU/mL
Source Murine Engelbreth-Holm-Swarm (EHS) tumor.
Content And Storage Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.