Learn More
R&D Systems™ CometAssay™ Single Cell Gel Electrophoresis Assay
Provides a simple and effective method for evaluating DNA damage in cells
Specifications
Quantity 50 Tests
View More Specs
Includes: Lysis Solution, 2 x 500mL; Comet LMAgarose, 15mL; Trevigen CometSlide, 25/kit; pH 10 200mM EDTA, 12.5mL; Kit with Silver Stain (Mfr. No. 4251-050-K) also includes four staining reagents and a fixation additive.
Description
Description
The principle of the assay is based upon the ability of denatured, cleaved DNA fragments to migrate out of the nucleoid under the influence of an electric field, whereas undamaged DNA migrates slower and remains within the confines of the nucleoid when a
Choose the Neutral CometAssay to detect double-stranded breaks or the Alkaline CometAssay for a more sensitive protocol used to detect smaller amounts of damage including single and double-stranded breaks
Cells are immobilized in a bed of low melting poi
Specifications
Specifications
50 Tests