Learn More
R&D Systems™ clasto-Lactacystin beta Lactone
Potent, highly specific, irreversible and cell-permeable inhibitor that covalently modifies the beta subunits of the proteasome
Description
Description
  • The beta-lactone intermediate is generated from the aqueous hydrolysis of lactacystin and is the active inhibitory species that reacts with the proteasome