missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Canine GM-CSF Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£399.56
Specifications
Gene ID (Entrez) | 403923 |
---|---|
Molecular Weight (g/mol) | 14.2 kDa |
Name | GM-CSF |
Quantity | 25μg |
Source | E. coli-derived canine GM-CSF protein Ala18-Lys144 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18381318
|
R&D Systems™
1546-GM-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£399.56
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18166970
|
R&D Systems™
1546-GM-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£399.56
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Canine GM-CSF Protein is derived from E. coli. The Recombinant Canine GM-CSF Protein has been validated for the following applications: Bioactivity.Specifications
403923 | |
GM-CSF | |
E. coli-derived canine GM-CSF protein Ala18-Lys144 | |
<0.01 EU per 1μg of the protein by the LAL method. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
14.2 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim | |
Recombinant |