missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Bovine IL-5 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£327.21
Specifications
Gene ID (Entrez) | 280825 |
---|---|
Molecular Weight (g/mol) | 14 kDa (monomer) |
Name | IL-5 |
Quantity | 25μg |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18329037
|
R&D Systems™
2525-BL-025 |
£327.21
25µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18157910
|
R&D Systems™
2525-BL-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£327.21
25µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Bovine IL-5 Protein is derived from NS0. The Recombinant Bovine IL-5 Protein has been validated for the following applications: Bioactivity.Specifications
280825 | |
IL-5 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
14 kDa (monomer) | |
25μg | |
Disulfide-linked homodimer | |
Recombinant |