missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Bovine Fibronectin Protein

Product Code. 18399237
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399237 5 mg 5mg
18370568 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18399237 Supplier R&D Systems™ Supplier No. 1030FN05M

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to support cell attachment and spreading when used as a substratum for cell culture. In this application, the recommended concentration for this effect is 1-5 μg/cm2. Fibronectin can also be added in the media to support cell spreading at a concentration of 0.5-50 μg/mL. Optimal concentration depends on cell type as well as the application or research objectives. Measured by the ability of the immobilized protein to support the adhesion of B16-F1 mouse melanoma cells. The ED50 for this effect is 10.0-100 ng/mL.
TRUSTED_SUSTAINABILITY

Specifikationer

Formulation Supplied as a 0.2μm filtered solution in Tris-HCl, NaCl and Urea.
Gene ID (Entrez) 280794
Name Fibronectin
Quantity 5 mg
Source Bovine plasma-derived Fibronectin protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. Avoid vortexing and excessive agitation.
Conjugate Unconjugated
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.