missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Avidin-Fluorescein

Product Code. 18164001
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18164001 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18164001 Supplier R&D Systems™ Supplier No. F0030

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications: Flow Cytometry

  • Optimized for use with monoclonal antibody-biotin reagents
  • Av-FITC is provided in a ready-to-use liquid format containing 0.15M NaCl, 25mM HEPES, 0.5% BSA, and 0.1% NaN3
  • FITC molecule can be excited by laser light at 488nm and has an optimal emission at wavelength at 525nm

Contains sodium azide as a preservative: sodium azide may react with lead and copper plumbing to form explosive metal azides. Flush with large volumes of water during disposal.

Applications:

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 Tests
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.