missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Camphor, TRC

Product Code. 30137089
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137089 10 g 10g
30136456 100 g 100g
30010746 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137089 Supplier Toronto Research Chemicals Supplier No. TRCC16580010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 464-49-3
Molecular Formula C10 H16 O
Molecular Weight (g/mol) 152.23
Synonym d-Camphor, (1R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, (+)-Camphor, Bicyclo[2.2.1]heptan-2-one, 1,7,7-trimethyl-, (1R,4R)- (9CI, ACI), (1R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one (ACI), Bicyclo[2.2.1]heptan-2-one, 1,7,7-trimethyl-, (1R)- (ZCI), Camphor, (1R,4R)-(+)- (8CI), (+)-(1R,4R)-Camphor, (+)-2-Bornanone, (+)-Camphor, (1R)-(+)-Camphor, (1R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, (1R)-Camphor, (1R,4R)-(+)-Camphor, (1R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, (R)-(+)-Camphor, (R)-Camphor, D-(+)-Camphor, D-Camphor, Alcanfor, Camphor, (+)-, d-Camphor, Dextrocamphora, Japanese camphor
IUPAC Name (1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1
Formula Weight 152.1201
Chemical Name or Material (+)-Camphor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.