missing translation for 'onlineSavingsMsg'
Learn More

(R)-BPO-27, MedChemExpress

Product Code. 16483847
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483847 50 mg 50mg
16457716 100 mg 100mg
16450368 5 mg 5mg
16421869 1 mg 1mg
16417908 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16483847 Supplier Medchem Express Supplier No. HY19778
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress (R)-BPO-27, the R enantiomer of BPO-27, is a potent, orally active and ATP-competitive CFTR inhibitor with an IC50 of 4 nM.

(R)-BPO-27, the R enantiomer of BPO-27, is a potent, orally active and ATP-competitive CFTR inhibitor with an IC50 of 4 nM.

Specifications

Chemical Name or Material (R)-BPO-27
CAS 1415390-47-4
Color White
Purity Grade Notes Research
Molecular Formula C26H18BrN3O6
Quantity 50 mg
Solubility Information DMSO : ≥ 14.28 mg/mL (26.04 mM)
SMILES O=C(C1=CC=C(O[C@@H](C2=CC=C(Br)O2)C3=C(N(C)C4=O)C(C(N4C)=O)=C(C5=CC=CC=C5)N36)C6=C1)O
Molecular Weight (g/mol) 548.34
Formula Weight 548.34
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.