missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-BINAP, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | (R)-BINAP |
| CAS | 76189-55-4 |
| Molecular Formula | C44H32P2 |
| Quantity | 250 mg |
| Synonym | 1,1'-[(1R)-[1,1'-Binaphthalene]-2,2'-diyl]bis[1,1-diphenyl-phosphine, (+)-(1R)-[1,1'-Binaphthalene]-2,2'-diylbis[diphenylphosphine], (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene |
| SMILES | c1ccc(cc1)P(c2ccccc2)c3ccc4ccccc4c3c5c(ccc6ccccc56)P(c7ccccc7)c8ccccc8 |
| Molecular Weight (g/mol) | 622.67 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction