missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-beta-Hydroxy-gamma-butyrolactone, 90+%, ee 99+%, Thermo Scientific™

Product Code. 11349116
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349116 1 g 1g
11369116 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349116 Supplier Thermo Scientific Alfa Aesar Supplier No. L20255.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-(+)-β-Hydroxy-γ-butyrolactone is used in the pharmaceutical industry as a chiral building block for the statin class of cholesterol-reducing drugs. It is also used to prepare HIV inhibitors and nutritional supplement such as L-carnitine.

Solubility
Miscible with ethanol and chloroform.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58081-05-3
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00211248
InChI Key FUDDLSHBRSNCBV-GSVOUGTGSA-N
Synonym r-4-hydroxydihydrofuran-2 3h-one, r-3-hydroxy-gamma-butyrolactone, 4r-4-hydroxyoxolan-2-one, r-+-beta-hydroxy-gamma-butyrolactone, 2 3h-furanone, dihydro-4-hydroxy-, 4r, pubchem6051, r-+-3-hydoxy-gama-butyro lactone, 4r-4-hydroxy-2-oxolanone, 4r-4-oxidanyloxolan-2-one, 3-hydroxybutyrolactone, 3r
PubChem CID 7060520
IUPAC Name (4R)-4-hydroxyoxolan-2-one
SMILES O[C@H]1COC(=O)C1

Specifications

Density 1.302
Boiling Point 98°C to 100°C (0.3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4655
Quantity 1 g
Beilstein 3648447
Solubility Information Miscible with ethanol and chloroform.
Optical Rotation +81° (c=2 in Methanol)
Formula Weight 102.09
Percent Purity ≥90%
Assay ee ≥99%
Chemical Name or Material (R)-(+)-beta-Hydroxy-gamma-butyrolactone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.