missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (R)-(+)-Bay K 8644

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
(R)-(+)-Bay K 8644 is a L-type Ca2+-channel blocker with negative inotropic and vasodilatatory effects in vivo. Enantiomer showing opposite effects to the racemate (±)-Bay K 8644 (Cat. No. 1544) and (S)-(-)- enantiomer (Cat. No. 1546). Also TMEM176B inhibitor. Induces IL-1β secretion and caspase-1 activation in bone marrow-derived dendritic cells in vitro and increases survival of tumor-bearing mice, both alone and in combination with immune checkpoint blockers.
Racemate and (S)-(-)-Enantiomer also available.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 356.3 |
| Purity | >97% |
| Molecular Weight (g/mol) | 356.301 |
| Chemical Name or Material | (R)-(+)-Bay K 8644 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction