missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Bay-K-8644, MedChemExpress

Product Code. p-200100106
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486318

Brand: Medchem Express HY15125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (R)-(+)-Bay-K-8644 is a calcium channel inhibitor. (R)-(+)-Bay-K-8644 inhibits Ba2+ currents (IBa) (IC50=975 nM).

(R)-(+)-Bay-K-8644 is a calcium channel inhibitor. (R)-(+)-Bay-K-8644 inhibits Ba2+ currents (IBa) (IC50=975 nM).

Specifications

Chemical Name or Material (R)-(+)-Bay-K-8644
CAS 98791-67-4
Color Deep Yellow
Purity Grade Notes Research
Molecular Formula C16H15F3N2O4
Quantity 10 mg
Solubility Information DMSO : ≥ 300 mg/mL (841.99 mM)
SMILES CC1=C([N+]([O-])=O)[C@H](C2=C(C(F)(F)F)C=CC=C2)C(C(OC)=O)=C(C)N1
Molecular Weight (g/mol) 356.3
Formula Weight 356.3
Percent Purity 96.6%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.