missing translation for 'onlineSavingsMsg'
Learn More

(R)-BAY-85-8501, MedChemExpress

Product Code. 16497746
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497746 100 mg 100mg
16493887 50 mg 50mg
16477958 10 mg 10mg
16410428 5 mg 5mg
16403716 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497746 Supplier Medchem Express Supplier No. HY19908B
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (R)-BAY-85-8501 is the less active Enantiomer of BAY-85-8501. BAY-85-8501 is a selective and potent inhibitor of Human Neutrophil Elastase (HNE), with an IC50 of 65 pM.

(R)-BAY-85-8501 is the less active Enantiomer of BAY-85-8501. BAY-85-8501 is a selective and potent inhibitor of Human Neutrophil Elastase (HNE), with an IC50 of 65 pM.

Specifications

Chemical Name or Material (R)-BAY-85-8501
CAS 2446175-39-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H17F3N4O3S
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (210.77 mM; Need ultrasonic)
SMILES N#CC1=C(C)N(C2=CC=CC(C(F)(F)F)=C2)C(N(C)[C@H]1C3=CC=C(C#N)C=C3S(=O)(C)=O)=O
Molecular Weight (g/mol) 474.46
Formula Weight 474.46
Percent Purity 98.57%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.