missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-alpha-Methylhistamine dihydrochloride, Thermo Scientific™

Product Code. 15444199
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15444199 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15444199 Supplier Thermo Scientific Alfa Aesar Supplier No. J61636.MC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 75614-89-0
Color White
Molecular Formula C6H13Cl2N3
MDL Number MFCD00083176
Quantity 100 mg
Sensitivity Hygroscopic
Synonym histamine, r--alpha-methyl-, dihydrochloride, r-1-1h-imidazol-4-yl propan-2-amine dihydrochloride, r--alpha-methyl histamine dihydrochloride, alpha-methylhistamine-r, r-alpha-methylhistamine dihydrochloride, r--, a-methylhistamine dihydrochloride, r---alpha-methylhistamine dihydrochloride, r---a-methyl-1h-imidazole-4-ethanamine dihydrobromide, a-methyl-1h-imidazole-4-ethanamine dihydrochloride, 2r-1-1h-imidazol-4-yl propan-2-amine dihydrochloride
InChI Key IZHCNQFUWDFPCW-ZJIMSODOSA-N
Optical Rotation −4° (c=0.8 in Water)
SMILES CC(CC1=CN=CN1)N.Cl.Cl
IUPAC Name (2R)-1-(1H-imidazol-5-yl)propan-2-amine;dihydrochloride
Molecular Weight (g/mol) 198.091
PubChem CID 11957567
Formula Weight 198.1
Physical Form Powder
Chemical Name or Material (R)-(-)-alpha-Methylhistamine dihydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.