missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha-Methylbenzyl isocyanate, TRC

Product Code. 30476154
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476154 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30476154 Supplier Toronto Research Chemicals Supplier No. TRCM2914101G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-(+)-alpha-Methylbenzyl isocyanate
CAS 33375-06-3
Molecular Formula C9H9NO
Quantity 1 g
Synonym (R)-(1-Isocyanatoethyl)-benzene, (+)-(R)-α-Phenethyl Isocyanate, (+)-1-Phenylethyl Isocyanate, (+)-α-Methylbenzyl Isocyanate, (+)-α-Phenylethyl Isocyanate, (R)-(+)-1-Phenethyl Isocyanate, (R)-(+)-Alpha-methylbenzyl Isocyanate, (R)-(+)-α-Methylbenzyl Isocyanate, (R)-(+)-α-Methylbenzyl Isocyanate, (R)-(1-Isocyanatoethyl)benzene, (R)-1-Phenylethyl Isocyanate, (R)-α-Methylbenzyl Isocyanate, (R)-α-Phenylethyl Isocyanate, R-(+)-1-Phenylethyl Isocyanate, [(1R)-1-Isocyanatoethyl]benzene
InChI Formula InChI=1S/C9H9NO/c1-8(10-7-11)9-5-3-2-4-6-9/h2-6,8H,1H3/t8-/m1/s1
SMILES C[C@@H](N=C=O)C1=CC=CC=C1
IUPAC Name (R)-(1-isocyanatoethyl)benzene
Molecular Weight (g/mol) 147.17
Formula Weight 147.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.