missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Alpha-Methoxyphenylacetic Acid, TRC

Product Code. 30474215
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474215 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474215 Supplier Toronto Research Chemicals Supplier No. TRCM26574010G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-(-)-alpha-Methoxyphenylacetic Acid
CAS 3966-32-3
Molecular Formula C9H10O3
Quantity 10 g
Synonym (R)-(-)-Methoxyphenyl-acetic Acid, (R)-alpha-Methoxy-benzeneacetic Acid, (-)-(R)-alpha-Methoxyphenylacetic Acid, (-)-(alphaR)-alpha-Methoxybenzeneacetic Acid, (-)-2-Methoxy-2-phenylacetic Acid, (-)-2-Methoxyphenylacetic Acid, (-)-Methoxyphenylacetic Acid, (-)-alpha-Methoxyphenylacetic Acid, (2R)-2-Methoxy-2-phenylacetic Acid, (2R)-2-Methoxy-2-phenylethanoic Acid, (2R)-O-Methylmandelic Acid, (R)-(-)-2-Methoxy-2-phenylacetic Acid, (R)-(-)-O-Methylmandelic Acid, (R)-(-)-alpha-Methoxyphenylacetic Acid, (R)-(-)-alpha-Methoxyphenylethanoic Acid, (R)-2-Methoxy-2-phenylacetic Acid, (R)-2-Methoxy-2-phenylacetic Acid, (R)-2-Methoxy-2-phenylethanoic Acid, (R)-2-Methoxyphenylacetic Acid, (R)-2-Phenyl-2-methoxyacetic Acid, (R)-Methoxy(phenyl)acetic Acid, (R)-Methoxyphenylacetic Acid, (R)-O-Methylmandelic Acid, (R)-alpha-Methoxy-alpha-phenylacetic Acid, (R)-alpha-Methoxy-alpha-phenylacetic Acid, (R)-alpha-Methoxybenzeneacetic Acid, (R)-alpha-Methoxybenzeneethanoic Acid, (R)-alpha-Methoxyphenylacetic Acid, (alphaR)-alpha-Methoxybenzeneacetic Acid, D-(-)-Methoxyphenylacetic Acid, D-alpha-Methoxyphenylacetic Acid
InChI Formula InChI=1S/C9H10O3/c1-12-8(9(10)11)7-5-3-2-4-6-7/h2-6,8H,1H3,(H,10,11)/t8-/m1/s1
SMILES CO[C@@H](C(=O)O)c1ccccc1
IUPAC Name (2R)-2-methoxy-2-phenylacetic acid
Molecular Weight (g/mol) 166.17
Formula Weight 166.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.