missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-alpha-Amino-gamma-butyrolactone hydrochloride monohydrate, 97%, Thermo Scientific™

Product Code. 11986191 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11986191

Brand: Thermo Scientific Acros 437570050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 104347-13-9
Molecular Formula C4H7NO2·ClH
Molecular Weight (g/mol) 155.59
InChI Key XBKCXPRYTLOQKS-AENDTGMFSA-N
Synonym d-homoserine lactone hydrochloride, r-+-alpha-amino-gamma-butyrolactone hydrochloride, r-3-aminodihydrofuran-2 3h-one hydrochloride, 3r-3-aminotetrahydrofuran-2-one hydrochloride, homoserine lactone hydrochloride, 2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r, r-+-a-amino-g-butyrolactone hydrochloride, pubchem14743, r-+-, a-amino-, a-butyrolactone hydrochloride, d-homoserine lactone hcl
PubChem CID 11954435
IUPAC Name (3R)-3-aminooxolan-2-one;hydrochloride
SMILES C1COC(=O)C1N.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 215.1°C to 217.1°C
Color Yellow to Brown
Elemental Analysis 29.64 to 32.12% C, 8.64 to 9.36% N
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Specific Rotation 30
Formula Weight 155.59
Percent Purity 97%
Physical Form Crystalline Powder, Powder, or Solid
Chemical Name or Material (R)-(+)-α-Amino-γ-butyrolactone hydrochloride monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-alpha-Amino-gamma-butyrolactone hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.