missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-alpha-Amino-gamma-butyrolactone hydrochloride monohydrate, 97%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Specifications
Specifications
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Melting Point | 215.1°C to 217.1°C |
| Color | Yellow to Brown |
| Elemental Analysis | 29.64 to 32.12% C, 8.64 to 9.36% N |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96.0 to max. 104% min. (Argentometry) |
| Packaging | Glass bottle |
| Quantity | 1 g |
| Specific Rotation | 30 |
| Show More |
Safety and Handling
Product Identifier
- (R)-(+)-alpha-Amino-gamma-butyrolactone hydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction